Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HECA Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531372
Description
Recommended positive controls: A549, HeLa. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes the homolog of the Drosophila headcase protein, a highly basic, cytoplasmic protein that regulates the re-entry of imaginal cells into the mitotic cycle during adult morphogenesis. In Drosophila, the encoded protein also inhibits terminal branching of neighboring cells during tracheal development.
Specifications
| HECA | |
| Polyclonal | |
| Unconjugated | |
| HECA | |
| dJ225E12.1; Gm869; H dJ225E12.1; HDC; hdc homolog, cell cycle regulator; HDCL; headcase homolog; headcase homolog (Drosophila); headcase protein homolog; HECA; hHDC; si:dkey-34f16.2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51696 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.7 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q9UBI9 | |
| HECA | |
| Recombinant protein encompassing a sequence within the C-terminus region of human HECA. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction