Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Heclin
GREENER_CHOICESDP

Catalog No. 543350 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
54-335-0 50 mg
54-331-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 54-335-0 Supplier R&D Systems™ Supplier No. 5433/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. HECT E3 ubiquitin ligase inhibitor

Heclin is a HECT E3 ubiquitin ligase inhibitor; inhibits Nedd4, Smurf2 and WWP1 (IC50 values are 6.3, 6.8 and 6.9 μM respectively). Selective for HECT-mediated over RING-mediated ubiquitination.

Specifications

Quantity 50 mg
Product Line Tocris
Product Type Heclin
Purity >98%
Molecular Weight (g/mol) 283.32
Inhibitors Heclin

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.