Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HECTD1 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP149926JF549
Description
HECTD1 Polyclonal antibody specifically detects HECTD1 in Human samples. It is validated for Western BlotSpecifications
HECTD1 | |
Polyclonal | |
Western Blot | |
E3 ligase for inhibin receptor, E3 ubiquitin-protein ligase HECTD1, EC 6.3.2.-, EULIR, HECT domain containing 1, HECT domain-containing protein 1, KIAA1131FLJ38315 | |
The immunogen for this product maps to a region between residue 2100 and 2150 of human HECT Domain Containing 1 using the numbering given in entry AAP13073.1 (GeneID 25831). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
25831 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction