Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HECTD1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP149926
Description
HECTD1 Polyclonal specifically detects HECTD1 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative).Specifications
HECTD1 | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
E3 ligase for inhibin receptor, E3 ubiquitin-protein ligase HECTD1, EC 6.3.2.-, EULIR, HECT domain containing 1, HECT domain-containing protein 1, KIAA1131FLJ38315 | |
Rabbit | |
Affinity Purified | |
RUO | |
25831 | |
Human | |
IgG |
Western Blot, Immunoprecipitation | |
1.0 mg/mL | |
Western Blot, Immunoprecipitation 2-5 μg/mg lysate, Western Blot (Negative) | |
Q9ULT8 | |
HECTD1 | |
The immunogen for this product maps to a region between residue 2100 and 2150 of human HECT Domain Containing 1 using the numbering given in entry AAP13073.1 (GeneID 25831). | |
100 μL | |
Primary | |
Based on 100% sequence identity, this antibody is predicted to react with X. tropicalis, Panda, Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, West Indian ocean coelacanth, Medaka fish and Spotted green pufferfish. | |
Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction