Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HECTD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103813
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103813 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103813 Supplier Invitrogen™ Supplier No. PA5103813
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total HECTD1.

Probable E3 ubiquitin-protein ligase which accepts ubiquitin from an E2 ubiquitin-conjugating enzyme in the form of a thioester and then directly transfers the ubiquitin to targeted substrates. May be required for development of the head mesenchyme and neural tube closure.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HECTD1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene HECTD1
Gene Accession No. Q69ZR2, Q9ULT8
Gene Alias A630086P08Rik; AI844876; b2b327Clo; E3 ligase for inhibin receptor; E3 ubiquitin-protein ligase HECTD1; EULIR; HECT domain containing 1; HECT domain containing E3 ubiquitin protein ligase 1; HECT domain E3 ubiquitin protein ligase 1; HECT domain-containing protein 1; HECTD1; HECT-type E3 ubiquitin transferase HECTD1; KIAA1131; opm; protein open mind; RGD1561653
Gene Symbols HECTD1
Host Species Rabbit
Immunogen A synthesized peptide derived from human HECTD1(Accession Q9ULT8), corresponding to amino acid residues L618-I668.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 207304, 25831, 362736
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.