Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HECTD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551778
Description
Immunogen sequence: RHWKLTGTNK SIRKNRNCSQ LIAAYKDFCE HGTKSGLNQG AISTLQSSDI LNLTKEQPQA KAGNGQNSCG VEDVLQLLRI LYIVASDPYS RISQEDGDEQ PQFTFPPDE Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 96%.
Probable E3 ubiquitin-protein ligase which accepts ubiquitin from an E2 ubiquitin-conjugating enzyme in the form of a thioester and then directly transfers the ubiquitin to targeted substrates. May be required for development of the head mesenchyme and neural tube closure.
Specifications
HECTD1 | |
Polyclonal | |
Unconjugated | |
HECTD1 | |
A630086P08Rik; AI844876; b2b327Clo; E3 ligase for inhibin receptor; E3 ubiquitin-protein ligase HECTD1; EULIR; HECT domain containing 1; HECT domain containing E3 ubiquitin protein ligase 1; HECT domain E3 ubiquitin protein ligase 1; HECT domain-containing protein 1; HECTD1; HECT-type E3 ubiquitin transferase HECTD1; KIAA1131; opm; protein open mind; RGD1561653 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
25831 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9ULT8 | |
HECTD1 | |
Recombinant protein corresponding to Human HECTD1. Recombinant protein control fragment (Product #RP-89077). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction