Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HECTD2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP216801
Description
HECTD2 Polyclonal specifically detects HECTD2 in Human samples. It is validated for Western Blot.Specifications
HECTD2 | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
EC 6.3.2.-, FLJ16050, FLJ37306, HECT domain containing 2, HECT domain-containing protein 2, probable E3 ubiquitin-protein ligase HECTD2 | |
Rabbit | |
88 kDa | |
0.1 mL | |
Primary | |
Human. Expected to cross react based on sequence identity: Bovine (90%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot | |
1mg/mL | |
Western Blot 1:500-1:3000 | |
Q5U5R9 | |
HECTD2 | |
Recombinant protein encompassing a sequence within the center region of human HECTD2. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
143279 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction