Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HECTD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558096
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558096 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558096 Supplier Invitrogen™ Supplier No. PA558096
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SPAHLVFPNI KNVREPPPIC LDVRQKQRTS MDASSSEMKA PVLPEPILPI QPKTVKDFQE DVEKVKSSGD WKA Highest antigen sequence identity to the following orthologs: Mouse - 67%, Rat - 71%.

Probable E3 ubiquitin-protein ligase which accepts ubiquitin from an E2 ubiquitin-conjugating enzyme in the form of a thioester and then directly transfers the ubiquitin to targeted substrates.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HECTD2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Hectd2
Gene Accession No. Q5U5R9
Gene Alias 4921524L07; A630025O09Rik; AW212605; HECT domain containing 2; HECT domain containing E3 ubiquitin protein ligase 2; HECT domain E3 ubiquitin protein ligase 2; HECT domain-containing protein 2; Hectd2; HECT-type E3 ubiquitin transferase HECTD2; hmm77879; Probable E3 ubiquitin-protein ligase HECTD2
Gene Symbols Hectd2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HECTD2. Recombinant protein control fragment (Product #RP-97141).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 143279
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.