Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HECTD2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569236
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 86%; Zebrafish: 100%.
Probable E3 ubiquitin-protein ligase which accepts ubiquitin from an E2 ubiquitin-conjugating enzyme in the form of a thioester and then directly transfers the ubiquitin to targeted substrates.
Specifications
HECTD2 | |
Polyclonal | |
Unconjugated | |
Hectd2 | |
4921524L07; A630025O09Rik; AW212605; HECT domain containing 2; HECT domain containing E3 ubiquitin protein ligase 2; HECT domain E3 ubiquitin protein ligase 2; HECT domain-containing protein 2; Hectd2; HECT-type E3 ubiquitin transferase HECTD2; hmm77879; Probable E3 ubiquitin-protein ligase HECTD2 | |
Rabbit | |
Affinity chromatography | |
RUO | |
143279 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q5U5R9 | |
Hectd2 | |
synthetic peptide directed towards the C terminal region of human HECTD2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction