Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HECTD4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559413
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559413 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559413 Supplier Invitrogen™ Supplier No. PA559413
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KETVHIPGAR CLYLRFDSRC SSQYDYDKLV IYAGPNTNSR KVAEYGGNTL GYGSRSVLGT GWPKDLVKVE GDTVTFSFEM RSGREHNTPD KAMWGFACTV R Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen HECTD4
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.30 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HECTD4
Gene Accession No. Q9Y4D8
Gene Alias A130010L24; AF-1 specific protein phosphatase; AU042671; BC019639; C12orf51; EG545802; Gm15800; HECT domain containing E3 ubiquitin protein ligase 4; HECT domain E3 ubiquitin protein ligase 4; HECT domain-containing protein 4; HECTD4; mKIAA0614; OTTMUSG00000026283; POTAGE; predicted gene 15800; probable E3 ubiquitin-protein ligase C12orf51; probable E3 ubiquitin-protein ligase HECTD4; transmembrane protein C12orf51
Gene Symbols HECTD4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HECTD4. Recombinant protein control fragment (Product #RP-97828).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283450
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.