Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HECW1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564914
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564914 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564914 Supplier Invitrogen™ Supplier No. PA564914
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: STEPESAQIQD SPMNNLMESG SGEPRSEAPE SSESWKPEQL GEGSVPDGPG NQSIELSRPA EEAAVITEAG DQGMVSVGPE GAGELLAQVQ KDIQPAPSAE ELAEQLDLGE EASALLLEDG EAPAS Highest antigen sequence identity to the following orthologs - mouse 45%, rat 44%.

E3 ubiquitin-protein ligase that mediates ubiquitination and subsequent degradation of DVL1. Also targets the mutant SOD1 protein involved in familial amyotrophic lateral sclerosis (FALS). Forms cytotoxic aggregates with DVL1, SSR3 and mutant SOD1 that lead to motor neuron death in FALS.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HECW1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HECW1
Gene Accession No. Q76N89
Gene Alias E3 ubiquitin-protein ligase HECW1; HECT type E3 ubiquitin ligase; HECT, C2 and WW domain containing E3 ubiquitin protein ligase 1; HECT, C2 and WW domain-containing protein 1; HECT-type E3 ubiquitin transferase HECW1; HECW1; hNEDL1; KIAA0322; NEDD4-like E3 ubiquitin-protein ligase 1; NEDD4-like ubiquitin-protein ligase 1; NEDL1; RGD1561038
Gene Symbols HECW1
Host Species Rabbit
Immunogen Recombinant Human HECW1. Recombinant protein control fragment (Product #RP-89679).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23072
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.