Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HECW1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552457
Description
Immunogen sequence: STEPESAQIQ DSPMNNLMES GSGEPRSEAP ESSESWKPEQ LGEGSVPDGP GNQSIELSRP AEEAAVITEA GDQGMVSVGP EGAGELLAQV QKDIQPAPSA EELAEQLDLG EEASALLLED GEAPAS Highest antigen sequence identity to the following orthologs: Mouse - 45%, Rat - 44%.
E3 ubiquitin-protein ligase that mediates ubiquitination and subsequent degradation of DVL1. Also targets the mutant SOD1 protein involved in familial amyotrophic lateral sclerosis (FALS). Forms cytotoxic aggregates with DVL1, SSR3 and mutant SOD1 that lead to motor neuron death in FALS.
Specifications
HECW1 | |
Polyclonal | |
Unconjugated | |
HECW1 | |
E3 ubiquitin-protein ligase HECW1; HECT type E3 ubiquitin ligase; HECT, C2 and WW domain containing E3 ubiquitin protein ligase 1; HECT, C2 and WW domain-containing protein 1; HECT-type E3 ubiquitin transferase HECW1; HECW1; hNEDL1; KIAA0322; NEDD4-like E3 ubiquitin-protein ligase 1; NEDD4-like ubiquitin-protein ligase 1; NEDL1; RGD1561038 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23072 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q76N89 | |
HECW1 | |
Recombinant protein corresponding to Human HECW1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction