Learn More
K1 Series Heelstop - Anti-slip Heel Traction Aid
Built with the toughest materials, the K1 Series Heelstop is made with ice-penetrating tungsten carbide spikes, TR freeze-resistant rubber, and reflective hi-viz strips.
Supplier:  K1 Series V6770170XL
 View more versions of this product
Catalog No. 19-060-824
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
  • Ice-penetrating tungsten carbide spikes cover the entire heel for maximum slip resistance
  • Freeze-resistant industrial quality TR rubber remains flexible even in the most extreme cold weather conditions
  • Reflective strips provide added safety when working in low lit working environments
  • Easy to attach and remove with convenient Velcro straps
  • Unique design allows driving and climbing without interference
Specifications
Specifications
Hook & Loop
Outdoor Surfaces, Ice Surfaces
X-Large
Heel Traction Aid
Unisex
Black
Vulcanized Rubber
Leather
Reusable
Yes
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.