Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

K1 Series Heelstop - Anti-slip Heel Traction Aid - Intrinsic

The K1 Series Heelstop is made with ice-penetrating tungsten carbide spikes, TR freeze-resistant rubber, and reflective hi-viz strips.

$57.50 - $59.40

Specifications

Closure Type Hook & Loop
Color Black
For Use With (Application) Outdoor Surfaces, Ice Surfaces
Material Vulcanized Rubber
Strap Material Leather
View More Specs

Products 4
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-060-819
View Documents
K1 Series
V5770170M
Medium
Pair for $57.50
Only null left
Add to Cart
 
19-060-818
View Documents
K1 Series
V5770170L
Large
Pair for $59.40
Only null left
Add to Cart
 
19-060-820
View Documents
K1 Series
V5770170XL
X-Large
Pair for $59.40
Only null left
Add to Cart
 
19-060-817
View Documents
K1 Series
V57701702XL
2X-Large
Pair for $57.50
Only null left
Add to Cart
 
Description

Description

  • Built with the toughest materials
  • Ice-penetrating tungsten carbide spikes cover the entire heel for maximum slip resistance
  • Intrinsic design is certified to prevent sparking when working in areas where combustible materials may be present
  • Proud to be the only fully certified ice-cleat brand in North America
  • Freeze-resistant industrial quality TR rubber remains flexible even in the most extreme cold weather conditions
  • Reflective strips provide added safety when working in low lit working environments
  • Easy to attach and remove with convenient Velcro straps
  • Unique design allows driving and climbing without interference
Specifications

Specifications

Hook & Loop
Outdoor Surfaces, Ice Surfaces
Leather
Unisex
Black
Vulcanized Rubber
Reusable
Yes
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.