Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HEF1 Monoclonal Antibody (2G9)
Mouse Monoclonal Antibody
$536.00 - $897.50
Specifications
Antigen | HEF1 |
---|---|
Clone | 2G9 |
Concentration | 1 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
01-674-895
|
Invitrogen™
MA15784 |
50 μL |
Each for $536.00
|
|
|||||
01-674-896
|
Invitrogen™
MA15785 |
100 μL |
Each for $897.50
|
|
|||||
Description
This antibody detects HEF1 in human, mouse, and rat samples. This antibody does not recognize p130cas. It has not been tested on Sin1. This antibody mostly detects HEF1 localized at the focal adhesion sites.
HEF-1 is a multifunctional protein involved in integrin-based signaling that affects cell motility, growth, apoptosis and oncogenic transformation. The Cas family of docking proteins have been the subject of intense research because of their role in cell motility, growth, apoptosis and oncogenic transformation. These proteins are substrates of focal adhesion kinase (FAK) and the Src family of tyrosine kinases two active targets for drug development. HEF1 protein production increases levels of mRNA transcripts that encode proteins associated with motility, cell transformation and invasiveness, including several metalloproteinases, MLCK, p160ROCK and ErbBi. HEF1 overproduction also mediates apoptosis in epithelial-derived cell lines, including MCF7 and HeLa cells. Recent clinical studies at another institution have found that overexpression of BCAR1 (p130Cas), a related protein, is associated with tamoxifen resistance. This highlights the importance of studying the role of this family of proteins in cancer prognosis.Specifications
HEF1 | |
1 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot | |
Unconjugated | |
Mouse | |
RUO | |
PBS with 0.1% sodium azide | |
O35177, Q14511 | |
18003, 291044, 4739 | |
Fusion protein, corresponding to amino acids 82-398 of Human HEF1. | |
Antibody |
2G9 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Monoclonal | |
Liquid | |
IgG1 κ | |
Human, Mouse, Rat | |
NEDD9 | |
Cas scaffolding protein family member 2; CAS2; CASL; CAS-L; cas-like docking; CASS2; Crk-associated substrate lymphocyte-specific protein; Crk-associated substrate related protein Cas-L; CRK-associated substrate-related protein; dJ49G10.2; dJ761I2.1; Enhancer of filamentation 1; Enhancer of filamentation 1 p55; HEF1; mEF1; Nedd9; NEDD-9; Neural precursor cell expressed developmentally down-regulated protein 9; neural precursor cell expressed, developmentally down-regulated 9; neural precursor cell expressed, developmentally down-regulated gene 9; p105; p130Cas-related protein; renal carcinoma antigen NY-REN-12 | |
NEDD9 | |
Primary | |
Protein G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title