Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HEF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111103
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111103 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111103 Supplier Invitrogen™ Supplier No. PA5111103
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: WWLCSLHGRQ GIVPGNRVKL LIGPMQETAS SHEQPASGLM QQTFGQQKLY QVPNPQAAPR DTIYQVPPSY QNQGIYQVPT GHGTQEQEVY QVPPS Highest antigen sequence indentity to the following orthologs: Mouse - 87%, Rat - 88%.

HEF-1 is a multifunctional protein involved in integrin-based signaling that affects cell motility, growth, apoptosis and oncogenic transformation. The Cas family of docking proteins have been the subject of intense research because of their role in cell motility, growth, apoptosis and oncogenic transformation. These proteins are substrates of focal adhesion kinase (FAK) and the Src family of tyrosine kinases two active targets for drug development. HEF1 protein production increases levels of mRNA transcripts that encode proteins associated with motility, cell transformation and invasiveness, including several metalloproteinases, MLCK, p160ROCK and ErbBi. HEF1 overproduction also mediates apoptosis in epithelial-derived cell lines, including MCF7 and HeLa cells. Recent clinical studies at another institution have found that overexpression of BCAR1 (p130Cas), a related protein, is associated with tamoxifen resistance. This highlights the importance of studying the role of this family of proteins in cancer prognosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HEF1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NEDD9
Gene Accession No. Q14511
Gene Alias Cas scaffolding protein family member 2; CAS2; CASL; CAS-L; cas-like docking; CASS2; Crk-associated substrate lymphocyte-specific protein; Crk-associated substrate related protein Cas-L; CRK-associated substrate-related protein; dJ49G10.2; dJ761I2.1; Enhancer of filamentation 1; Enhancer of filamentation 1 p55; HEF1; mEF1; Nedd9; NEDD-9; Neural precursor cell expressed developmentally down-regulated protein 9; neural precursor cell expressed, developmentally down-regulated 9; neural precursor cell expressed, developmentally down-regulated gene 9; p105; p130Cas-related protein; renal carcinoma antigen NY-REN-12
Gene Symbols NEDD9
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human HEF1. Recombinant protein control fragment (Product #RP-102849).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4739
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.