Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HEG Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA572634
Description
HEG Polyclonal Antibody for Western Blot
Receptor component of the CCM signaling pathway which is a crucial regulator of heart and vessel formation and integrity May act through the stabilization of endothelial cell junctions. [UniProt].
Specifications
HEG | |
Polyclonal | |
Unconjugated | |
HEG1 | |
4632417D23Rik; 5530401I02Rik; 9530025L16Rik; BC038614; Gm629; heart development protein with EGF like domains 1; heart development protein with EGF-like domains 1; heart of glass; heart of glass soluble isoform 1; heart of glass soluble isoform 2; heart of glass transmembrane isoform; HEG; HEG homolog 1; HEG homolog 1 (zebrafish); Heg_SE1; Heg_SE2; Heg_TM; HEG1; KIAA1237; MST112; MSTP112; OTTMUSG00000027315; protein HEG; protein HEG homolog 1; RGD1564641 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
406202 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
0 | |
HEG1 | |
KLH conjugated synthetic peptide between 691-717 amino acids from the C-terminal region of DANRE heg. | |
400 μL | |
Primary | |
Zebrafish | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction