Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HEG1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552842
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552842 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552842 Supplier Invitrogen™ Supplier No. PA552842
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NQEDFSTVSS KEGVMVQTSG KSHAASDAPE NLTLLAETAD ARGRSGSSSR TNFTILPVGY SLEIATALTS QSGNLASESL HLPSSSSEFD ERIAAFQTKS GTASEMGTER AMGLSEEWTV Highest antigen sequence identity to the following orthologs: Mouse - 43%, Rat - 38%.

Receptor component of the CCM signaling pathway which is a crucial regulator of heart and vessel formation and integrity May act through the stabilization of endothelial cell junctions. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen HEG1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HEG1
Gene Accession No. Q9ULI3
Gene Alias 4632417D23Rik; 5530401I02Rik; 9530025L16Rik; BC038614; Gm629; heart development protein with EGF like domains 1; heart development protein with EGF-like domains 1; heart of glass; heart of glass soluble isoform 1; heart of glass soluble isoform 2; heart of glass transmembrane isoform; HEG; HEG homolog 1; HEG homolog 1 (zebrafish); Heg_SE1; Heg_SE2; Heg_TM; HEG1; KIAA1237; MST112; MSTP112; OTTMUSG00000027315; protein HEG; protein HEG homolog 1; RGD1564641
Gene Symbols HEG1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HEG1. Recombinant protein control fragment (Product #RP-90450).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57493
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.