Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HEG1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983777
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3777 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3777 Supplier Bioss Supplier No. BS15449R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Receptor component of the CCM signaling pathway which is a crucial regulator of heart and vessel formation and integrity. May act through the stabilization of endothelial cell junctions.

Specifications

Antigen HEG1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene HEG1
Gene Accession No. Q9ULI3
Gene Alias heart development protein with EGF like domains 1; heart development protein with EGF-like domains 1; heart of glass; heart of glass soluble isoform 1; heart of glass soluble isoform 2; heart of glass transmembrane isoform; heg; HEG homolog 1; Heg_SE1; Heg_SE2; Heg_TM; HEG1; KIAA1237; MST112; MSTP112; protein HEG; Protein HEG homolog 1
Gene Symbols HEG1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human HEG1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57493
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.