Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heidolph™ Hei-PLATE Mix 'n' Heat Magnetic Stirring Hotplate Accessory, Protective Cover
SDP

Catalog No. p-7239920 Shop All Heidolph North America Products
Click to view available options
For Use With (Equipment):
Hei-PLATE Mix 'n' Heat
Hei-PLATE Mix 20 L
This item is not returnable. View return policy

Catalog No. 13-880-269

Supplier: Heidolph™ 036306041

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Use this Silicone Protective Cover for Hei-PLATE Magnetic Stirrers.

  • Replaceable cover against splashes and dripping water

    Specifications

    Color Silver/White
    For Use With (Equipment) Hei-PLATE Mix 20 L
    Type Silicone Protective Cover
    Material Silicone
    WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Your feedback has been submitted: Thank you for helping us improve our website.