Learn More
Heidolph™ Hei-Vap Rotary Evaporator Accessories - Glassware Sets
SDP
Accessories for Hei-VAP rotary evaporator
Supplier:  Heidolph™ 036301790
 View more versions of this product
Catalog No. 13-878-636
Only null left
Add to Cart
Request a Quote
Description
Description
  • Glassware set includes one 1000 mL evaporating flask and one 1000 mL receiving flask
  • All glassware sets feature GL 10 thread; the inlet tube can be connected with a threaded fitting
  • All glassware sets are also available with transparent plastic halar coating for G5B and abcite coating for G1B/G3B/G6B
  • Glassware sets G3 and G6 allow for the automated distillation program AUTOaccurate
Specifications
Specifications
Glassware G5 Set
Hei-VAP Rotary Evaporator
Dry ice condenser, 1000 mL NS 29/32 Joint sized evaporating flask, 1000 mL receiving flask
NS 29/32
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.