Learn More
Heidolph™ Hei-Vap Rotary Evaporator Accessories - Glassware Sets
GSA_VASDP
Accessories for Hei-VAP rotary evaporator
$2708.90 - $8652.00
Specifications
For Use With (Equipment) Hei-VAP Rotary Evaporator
View More Specs
Products 9
Catalog Number Mfr. No. Type Taper Price Quantity & Availability  
Catalog Number Mfr. No. Type Taper Price Quantity & Availability  
13-878-579
SDP
View Documents
Heidolph™
036301220
Glassware G1 Set NS 29/32
Each for $2,708.90
Only null left
Add to Cart
13-878-597
SDP
View Documents
Heidolph™
036301400
Glassware G3 Set NS 29/32
Each for $3,120.90
Only null left
Add to Cart
13-878-580
SDP
View Documents
Heidolph™
036301230
Glassware G1 Set NS 24/40
Each for $2,801.60
Only null left
Add to Cart
13-878-636
SDP
View Documents
Heidolph™
036301790
Glassware G5 Set NS 29/32
Each for $6,571.40
Only null left
Add to Cart
13-878-582
SDP
View Documents
Heidolph™
036301250
Glassware G1B Set NS 24/40
Each for $3,223.90
Only null left
Add to Cart
13-878-600
SDP
View Documents
Heidolph™
036301430
Glassware G3B Set NS 24/40
Each for $4,738.00
Only null left
Add to Cart
13-878-661
GSA_VASDP
View Documents
Heidolph™
036302040
Glassware G6 Set 24/40
Each for $7,539.60
Only null left
Add to Cart
13-878-663
GSA_VASDP
View Documents
Heidolph™
036302060
Glassware G6B Set 24/40
Each for $8,652.00
Only null left
Add to Cart
13-878-800
SDP
View Documents
Heidolph™
036301280CA
Glassware G3 Set NS 24/40 N/A N/A N/A
Description
Description
  • Glassware set includes one 1000 mL evaporating flask and one 1000 mL receiving flask
  • All glassware sets feature GL 10 thread; the inlet tube can be connected with a threaded fitting
  • All glassware sets are also available with transparent plastic halar coating for G5B and abcite coating for G1B/G3B/G6B
  • Glassware sets G3 and G6 allow for the automated distillation program AUTOaccurate
Specifications
Specifications
Hei-VAP Rotary Evaporator
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.