Learn More
Heidolph™ Hei-Vap Rotary Evaporator Accessories - Glassware Sets
Accessories for Hei-VAP rotary evaporator
Supplier: Heidolph™ 036302040
Description
- Glassware set includes one 1000 mL evaporating flask and one 1000 mL receiving flask
- All glassware sets feature GL 10 thread; the inlet tube can be connected with a threaded fitting
- All glassware sets are also available with transparent plastic halar coating for G5B and abcite coating for G1B/G3B/G6B
- Glassware sets G3 and G6 allow for the automated distillation program AUTOaccurate
Specifications
Glassware G6 Set | |
Hei-VAP Rotary Evaporator | |
Vertical condenser, centerpiece with valve, 1000 mL 24/40 standard ground joint evaporating flask, 1000 mL receiving flask | |
24/40 |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov