Learn More
Heidolph™ Hei-Vap™ Rotary Evaporator Accessories - Receiving Flasks
SDP
Coated or standard receiving flasks
$204.87 - $852.84
Specifications
For Use With (Equipment) Hei-VAP Rotary Evaporator
Taper 35/20
View More Specs
Products 11
Catalog Number Mfr. No. Volume (Metric) Type Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Type Price Quantity & Availability  
13-878-707
SDP
View Documents
Heidolph™
036302500
250 mL Standard Receiving Flask
Each for $219.91
Only null left
Add to Cart
13-878-714
SDP
View Documents
Heidolph™
036302570
2000 mL Standard Receiving Flask
Each for $442.39
Only null left
Add to Cart
13-878-575
SDP
View Documents
Heidolph™
036301180
1000 mL Coated Receiving Flask
Each for $777.65
Only null left
Add to Cart
13-878-706
SDP
View Documents
Heidolph™
036302490
100 mL Coated Receiving Flask
Each for $307.97
Only null left
Add to Cart
13-878-716
SDP
View Documents
Heidolph™
036302590
3000 mL Standard Receiving Flask
Each for $799.28
Only null left
Add to Cart
13-878-710
SDP
View Documents
Heidolph™
036302530
500 mL Coated Receiving Flask
Each for $361.53
Only null left
Add to Cart
13-878-715
SDP
View Documents
Heidolph™
036302580
2000 mL Coated Receiving Flask
Each for $777.65
Only null left
Add to Cart
13-878-705
SDP
View Documents
Heidolph™
036302480
100 mL Standard Receiving Flask
Each for $204.87
Only null left
Add to Cart
13-878-574
SDP
View Documents
Heidolph™
036301170
1000 mL Standard Receiving Flask
Each for $246.69
Only null left
Add to Cart
13-878-717
SDP
View Documents
Heidolph™
036302600
3000 mL Coated Receiving Flask
Each for $852.84
Only null left
Add to Cart
13-878-708
SDP
View Documents
Heidolph™
036302510
250 mL Coated Receiving Flask
Each for $307.97
Only null left
Add to Cart
Description
Description
Standard and Surlyn-coated receiving flasks for Heidolph Hei-VAP Rotary Evaporator
  • NS 35/20 joint size
Specifications
Specifications
Hei-VAP Rotary Evaporator
35/20
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.