Learn More
Heidolph™ Hei-Vap™ Rotary Evaporator Accessories - Foam Brakes
For use with Heidolph Hei-VAP series rotary evaporators
$637.06
Specifications
| Type | Foam Brake |
|---|---|
| For Use With (Equipment) | Hei-VAP Rotary Evaporator |
| Includes | Flask clamp |
| Catalog Number | Mfr. No. | Taper | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Taper | Price | Quantity & Availability | |||||
13-878-703
![]() |
Heidolph™
036302460 |
29/32 |
Each for $637.06
|
|
|||||
13-878-704
![]() |
Heidolph™
036302470 |
24/40 |
Each for $637.06
|
|
|||||
Description
- The foam brake prevents contents of the evaporating flask from being drawn into the condenser in the event of bumping.
Specifications
| Foam Brake | |
| Hei-VAP Rotary Evaporator | |
| Flask clamp |