Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Heidolph™ Hei-Vap™ Rotary Evaporator Accessories - Spiders

For use with Heidolph Hei-VAP series rotary evaporators
Supplier: Heidolph™ 036302610
Description
- Processes multiple samples at the same time
- Choice of models: NS 14.5 spider with 6, 12 or 20 distillation sleeves (20 mL) or NS 24 spider with 5 flasks (50 or 100 mL)
- Available in NS 24/40 or 29/32 joint size
Specifications
| Spider Flasks | |
| 6 x 20 mL Distillation Sleeves | |
| NS 14.5 |
| Hei-VAP Rotary Evaporator | |
| 24/40 | |
| 20 mL |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction