Learn More
Heidolph™ Hei-VAP Industrial and Laborota 20 Rotary Evaporator Accessories
SDP
Catalog No. p-4510946 Shop All Heidolph North America Products
Click to view available options
Type:
Tube Set
Vapor Temperature Sensor
Catalog No. 13-878-860
Supplier: Heidolph™ 036303080
Only null left
Add to Cart
Add to Cart
Accessories for use with Heidolph Hei-VAP Industrial and Laborota 20 rotary evaporators only
  • The vacuum sensor controls and displays the actual vacuum controls in addition the maximum permissible pressure.
  • The vacuum valve controls vacuum.
  • Vapor temperature sensor measure the temperature of vapors in the condenser.
  • Tubing set is 6 meter of vacuum and 10 meter of enforced cooling tube including hose clamps.
Specifications
Type Vapor Temperature Sensor
For Use With (Equipment) Hei-VAP Industrial and Laborota 20 Rotary Evaporator
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.