Learn More
Heidolph™ Rotary Evaporator Accessories - Interchangeable Glassware
Shop All Heidolph North America Products
Click to view available options
Type:
1000 mL Evaporating Flask
Clamp S 35
G1 Diagonal Condenser with safety coating
G3 Vertical Condenser
Jacket for Condensate Trap, G5 Condenser
Description
- All glassware can be interchanged among rotary evaporator models
- Each set includes one 1000 mL evaporating flask and one 1000 mL receiving flask
- All sets are available with standard or plastic safety-coated glassware (halar coating for G5B and abcite coating for G1B/G3B/G6B)
- Evaporating flasks and vapor tubes with the standard ground joint NS 24/40 are also available with NS 29/32 ground joints
Specifications
Specifications
| Type | 1000 mL Evaporating Flask |
| For Use With (Equipment) | Rotary Evaporator |
| Taper | 29/32 |
| Volume (Metric) | 1000 mL |