Learn More
Heidolph™ Rotary Evaporator Accessories - Interchangeable Glassware
SDP
Five condenser assemblies allow customization of evaporator glassware for any application
Supplier:  Heidolph™ 036301390
 View more versions of this product
Catalog No. 13-878-596
Only null left
Add to Cart
Description
Description
  • All glassware can be interchanged among rotary evaporator models
  • Each set includes one 1000 mL evaporating flask and one 1000 mL receiving flask
  • All sets are available with standard or plastic safety-coated glassware (halar coating for G5B and abcite coating for G1B/G3B/G6B)
  • Evaporating flasks and vapor tubes with the standard ground joint NS 24/40 are also available with NS 29/32 ground joints
Specifications
Specifications
Support for G3, G4, G6 Condensers
Rotary Evaporator
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.