Learn More
Heidolph™ Rotary Evaporator Accessories - Interchangeable Glassware
SDP
Catalog No. p-4379395 Shop All Heidolph North America Products
Click to view available options
Type:
1000 mL Evaporating Flask
Clamp S 35
G1 Diagonal Condenser with safety coating
G3 Vertical Condenser
Jacket for Condensate Trap, G5 Condenser
Catalog No. 13-878-570
Supplier: Heidolph™ 036301130
Only null left
Add to Cart
Add to Cart
Five condenser assemblies allow customization of evaporator glassware for any application
  • All glassware can be interchanged among rotary evaporator models
  • Each set includes one 1000 mL evaporating flask and one 1000 mL receiving flask
  • All sets are available with standard or plastic safety-coated glassware (halar coating for G5B and abcite coating for G1B/G3B/G6B)
  • Evaporating flasks and vapor tubes with the standard ground joint NS 24/40 are also available with NS 29/32 ground joints
Specifications
Type G1 Diagonal Condenser with safety coating
For Use With (Equipment) Rotary Evaporator
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.