Learn More
Heidolph™ Radleys Enlarging Adapter and Reducing Adapter
Shop All Heidolph North America Products
Click to view available options
Thread Size:
B24 to B19
B24 to B29
B29 to B19
B29 to B24
B29 to B34
B34 to B19
B34 to B24
B34 to B29
Description
- Features Rodaviss joint and is compatible with Reactor-Ready lids
Specifications
Specifications
| Product Type | Enlarging Adapter |
| Standard Taper Joint No. | B24 to B29 |
| Thread Size | B24 to B29 |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov