Learn More
Heidolph™ Radleys™ Reaction Vessel Accessory: DN200 Lids
Shop All Heidolph North America Products
Click to view available options
Type:
6-Neck Lid
7-Neck Lid
Description
- For reaction vessels used in Reactor-Ready™ Pilot Lab Reactor
- Lid with grease free Rodaviss joints that are interchangeable with ordinary 1-in-10 B and A length tapered conical joints and will not jam
- Available as 6 or 7 neck
Inlets in 6 Neck DN200 Lid
- 1 x B34 central vertical - stirrer
- 1 x B19 side vertical - probe
- 1 x B24 side vertical - gas purge
- 1 x B29 side vertical - dropping funnel, bubbler
- 1 x B34 side vertical - condenser and reflux
- 1 x B45 side angled - solid additions
Inlets in 7 Neck DN200 Lid
- 1 x B34 central vertical - stirrer
- 1 x B19 side vertical - probe
- 1 x B24 side vertical - gas purge
- 1 x B29 side vertical - drop funnel, bubbler
- 2 x B34 side vertical - condenser and reflux
- 1 x B45 side angled - solid additions
Specifications
Specifications
| Type | 6-Neck Lid |
| For Use With (Equipment) | Reaction Vessel |