Learn More
Heidolph™ Radleys Rotary Evaporator Adapters
Rotary evaporator adapters for use with Carousel 6 Plus Reaction Station™
Supplier: Heidolph™ 015880620
Description
Choice of size Rodaviss B29 socket to B24 cone, and 24/40 socket to B24 cone.
Specifications
24/40 Socket to B24 Cone | |
Rotary Evaporator Adapter | |
Carousel 6 Plus Reaction Station™ |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov