Learn More
Heidolph™ StarFish™ Work Station Accessories - Support Rods
SDP
Catalog No. p-4889032 Shop All Heidolph North America Products
Click to view available options
Type:
Support Rod
Support Split Rod
Accessories for use with Heidolph StarFish™ Work Station
  • Stainless steel 12 mm diameter x 650 mm long.
  • Rods screw directly into base plate.
  • Specifications
    For Use With (Equipment) Heidolph StarFish Work Station
    Type Support Split Rod
    WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
    Product Content Correction
    Your input is important to us. Please complete this form to provide feedback related to the content on this product.
    Product Title
    Your feedback has been submitted: Thank you for helping us improve our website.