Learn More
Heidolph™ Stirrer Chucks for RZR Overhead Stirrer
Shop All Heidolph North America Products
Click to view available options
For Use With (Equipment):
2102 Z Stirrer
RZR 1 Stirrer
RZR 2020 - 2102 Stirrers
Description
13880150
- For use with RZR 1 stirrer
- 8 mm
- For use with RZR 2020 - 2102 stirrers
- 10.5 mm
- For use with 2102 Z stirrer
- 10.5 mm
Specifications
Specifications
For Use With (Equipment) | RZR 1 Stirrer |
Type | Stirrer Chuck |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov