Learn More
Description
Specifications
Specifications
| Quantity | 2 Vials |
| Storage Buffer | The protein lysate is prepared in 1X Laemmli sample buffer with BME. Boiling before loading is not necessary. |
| Cellular Fraction | Hypoxic / Normoxic |
| Host Species | Human |
| Content And Storage | Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. |
| Product Line | Novus |
| For Use With (Application) | SDS-Page |
| Tissue | Cervix |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
