Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HELB Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA541297
Description
Peptide sequence: FGRFPITGAW WRVKVQVKPV VGSRSYQYQV QGFPSYFLQS DMSPPNQKHI Sequence homology: Cow: 86%; Guinea Pig: 79%; Human: 100%; Pig: 86%; Rat: 86%.
HELB gene ontology annotations related to this gene include cellular response to DNA damage stimulus; DNA replication, synthesis of RNA primer; negative regulation of double-strand break repair via homologous recombination; regulation of DNA double-strand break processing.
Specifications
HELB | |
Polyclonal | |
Unconjugated | |
HELB | |
AI447783; D10Ertd664e; DHB; DNA helicase B; hDHB; HELB; helicase (DNA) B; RGD1560103 | |
Rabbit | |
Protein A | |
RUO | |
92797 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8NG08 | |
HELB | |
Synthetic peptide directed towards the N-terminal of human HELB (aa 77-126). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction