Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HELB Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144788
Description
Immunogen sequence: VDNATRISRR QFPKFDAELN ISDNPTLPIS IQDKTFIFVR LPEEDASSQS SKTNHHSCLY SAVKTLLQEN NLQNAKTSQF IAFRRQDCDL INDCCC Antigen sequence identity: Mouse 80%; Rat 79%.
HELB gene ontology annotations related to this gene include cellular response to DNA damage stimulus; DNA replication, synthesis of RNA primer; negative regulation of double-strand break repair via homologous recombination; regulation of DNA double-strand break processing.
Specifications
HELB | |
Polyclonal | |
Unconjugated | |
HELB | |
AI447783; D10Ertd664e; DHB; DNA helicase B; hDHB; HELB; helicase (DNA) B; RGD1560103 | |
Rabbit | |
Affinity chromatography | |
RUO | |
92797 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
IgG |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NG08 | |
HELB | |
Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 662-757. Recombinant protein control fragment (Product #RP-110101). | |
100 μL | |
Primary | |
Human | |
Antibody | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction