Learn More
Helena Laboratories™ ColoCARE™ FOB Test Kit
Description
- CLIA Complexity: Waived
- Throw-in-the-bowl test for detecting pre-symptomatic occult bleeding caused by gastrointestinal diseases
- Clean and disposable, easy for elderly patients to see and interpret, and extremely sensitive, with no increase in the false positive rate
- Requires no stool handling, no chemical developers, no laboratory processing, and no mailing of biohazards
- Test pad consists of biodegradable paper chemically treated with a chromogen; floated on the water surface in the toilet bowl
- If detectable blood is present, the hemoglobin reacts with the chromogen, and a blue and/or green color reaction occurs
- With three reaction sites: a large test square and two smaller control squares to verify the system functions properly
Specifications
Specifications
| Clia Complexity | Waived |
| Type | Office Pack |
| For Use With (Application) | To test fecal occult blood |
| Includes | 50 Patient kits, each containing 3 ColoCARE Tests, 1 patient instruction sheet, 1 ColoCARE reply card, 50 “ColoCARE Dispensed Today” labels, instructions |
| Quantity | 50 Pack |