Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Helicin, 99%, Thermo Scientific™

Catalog No. ac226050010
Click to view available options
Quantity:
1 g

Chemical Identifiers

CAS 618-65-5
Molecular Formula C13H16O7
Molecular Weight (g/mol) 284.26
MDL Number MFCD00006589
InChI Key BGOFCVIGEYGEOF-UJPOAAIJSA-N
Synonym helicin, 2-beta-d-glucopyranosyloxy benzaldehyde, unii-6n4ti8p9r5, 2-formylphenyl beta-d-glucopyranoside, salicylaldehyde beta-d-glucopyranoside, helicine, salicylaldehyde b-d-glucoside, salicylaldehyde, a-d-glucoside, salicylaldehyde-beta-d-glucoside
PubChem CID 101799
ChEBI CHEBI:73235
IUPAC Name 2-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxybenzaldehyde
SMILES OC[C@H]1O[C@@H](OC2=CC=CC=C2C=O)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

Melting Point 178°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Beilstein 31, 223
Merck Index 15, 4663
Specific Rotation Condition −60° (20°C c=2,H2O)
Specific Rotation -60
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol
Formula Weight 284.26
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Helicin, 99%
Show More Show Less
EINECSNumber 210-558-9

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.