Learn More
Invitrogen™ Helicobacter pylori 19 kDa Monoclonal Antibody (A108H)

Description
The MA1-7456 antibody reacts with Helicobacter pylori 19 kDa Ag in bacterial and infected tissue samples. MA1-7456 has been successfully used in ELISA, Immunofluorescence and Western blot applications. The MA1-7456 antibody was raised against H. pylori 19 kDa antigen.
Specifications
Specifications
| Antigen | Helicobacter pylori 19 kDa |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | A108H |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.2 |
| Gene Alias | H. pylori |
| Host Species | Mouse |
| Immunogen | H. pylori 19 kDa antigen. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.