Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Helicobacter pylori 54 kDa Monoclonal Antibody (G56H)
GREENER_CHOICE

Catalog No. PIMA17457
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA17457 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA17457 Supplier Invitrogen™ Supplier No. MA17457
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The MA1-7457 antibody reacts with Helicobacter pylori 54 kDa (flagellin) Ag in bacterial and infected tissue samples. MA1-7457 has been successfully used in ELISA, Immunofluorescence and Western blot applications. The MA1-7457 antibody was raised against H. pylori 54 kDa antigen.

Helicobacter Pylori (H. Pylori) is known to cause peptic ulcers and chronic gastritis in human. It is associated with duodenal ulcers and may be involved in development of adenocarcimona and low grade lymphoma of mucosa associated lymphoid tissue in the stomach. It is a microaerophilic gram-Negative bacterium, helix-shaped, and with many diverse strains.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Helicobacter pylori 54 kDa
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone G56H
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias H. pylori
Host Species Mouse
Immunogen H. pylori 54 kDa antigen.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.