Learn More
Invitrogen™ Helicobacter pylori 58 kDa Monoclonal Antibody (C81F)

Description
The MA1-7459 antibody reacts with Helicobacter pylori 58 kDa Ag in bacterial and infected tissue samples. MA1-7459 has been successfully used in ELISA, Immunofluorescence and Western blot applications. The MA1-7459 antibody was raised against H. pylori 58 kDa antigen.
Specifications
Specifications
| Antigen | Helicobacter pylori 58 kDa |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | C81F |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.2 |
| Gene Alias | H. pylori |
| Host Species | Mouse |
| Immunogen | H. pylori 58 kDa antigen. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.