Learn More
Invitrogen™ Helicobacter pylori 19 kDa Monoclonal Antibody (G14F)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA17460
Description
The MA1-7460 antibody reacts with Helicobacter pylori 19 kDa (weak for 58 kDa) Ag in bacterial and infected tissue samples. MA1-7460 has been successfully used in ELISA, Immunofluorescence and Western blot applications. The MA1-7460 antibody was raised against H. pylori 19 kDa antigen.
Helicobacter Pylori (H. Pylori) is known to cause peptic ulcers and chronic gastritis in human. It is associated with duodenal ulcers and may be involved in development of adenocarcimona and low grade lymphoma of mucosa associated lymphoid tissue in the stomach. It is a microaerophilic gram-Negative bacterium, helix-shaped, and with many diverse strains.
Specifications
Helicobacter pylori 19 kDa | |
Monoclonal | |
0.1 mg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Mouse | |
Protein A | |
RUO | |
Bacteria | |
Antibody | |
IgG1 |
ELISA, Western Blot, Immunocytochemistry | |
G14F | |
Unconjugated | |
H. pylori | |
H. pylori 19 kDa antigen. | |
100 μg | |
Primary | |
4°C | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.