Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Helicobacter Pylori Antibody (B-53.4), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311433V
Description
Helicobacter Pylori Monoclonal antibody specifically detects Helicobacter Pylori in H. pylori samples. It is validated for Flow Cytometry, ELISA, ImmunofluorescenceSpecifications
Helicobacter Pylori | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
H. pylori | |
Purified |
Flow Cytometry, ELISA, Immunofluorescence | |
B-53.4 | |
Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence | |
H. pylori | |
Total sonicate of Helicobacter pylori | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction