Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Helicobacter Pylori Antibody (HP/1336), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308499F
Description
Helicobacter Pylori Monoclonal antibody specifically detects Helicobacter Pylori in H. pylori samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/ ImmunofluorescenceSpecifications
Helicobacter Pylori | |
Monoclonal | |
FITC | |
H. pylori | |
Total sonicate of Helicobacter pylori | |
100 μg | |
Primary | |
Store at 4°C in the dark. | |
IgG1 κ |
Flow Cytometry, ELISA, Immunofluorescence | |
HP/1336 | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
H. pylori | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction