Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Helicobacter Pylori Antibody (HP/212), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2452090.2MG
Description
Ensure accurate, reproducible results in ELISA, Flow Cytometry, Immunofluorescence
Helicobacter Pylori Monoclonal antibody specifically detects Helicobacter Pylori in Bacteria samples. It is validated for Flow Cytometry, Immunofluorescence.Specifications
Helicobacter Pylori | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunofluorescence 1 - 2 ug/ml | |
H. pylori | |
Total sonicate of Helicobacter pylori | |
0.2 mg | |
Primary | |
Bacteria | |
Purified |
Flow Cytometry, Immunofluorescence | |
HP/212 | |
Unconjugated | |
10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
Mouse | |
Protein A or G purified | |
RUO | |
The spiral shaped bacterium Helicobacter pyloriis strongly associated with inflammation of the stomach and is also implicated in the development of gastric malignancy. H. pyloriis known to cause peptic ulcers and chronic gastritis in human. It is associated with duodenal ulcers and may be involved in development of adenocarcinoma and low-grade lymphoma of mucosa associated lymphoid tissue in the stomach. This antibody stains the individual H. pylori bacterium when it presents on the surface of the epithelium or in the cytoplasm of the epithelial cells in biopsy tissue sections from the antrum and body of the stomach. | |
Store at 4C. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction