Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Helicobacter Pylori HP-NAP Recombinant Rabbit Monoclonal Antibody (RM413)
GREENER_CHOICE

Catalog No. PIMA544570
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544570 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA544570 Supplier Invitrogen™ Supplier No. MA544570
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This antibody reacts to Histone H2A.X acetylated at Lysine 5 (K5ac). No cross reactivity with non-modified Lysine 5 or other acetylated Lysines in histone H2A.

In H. pylori, the Helicobacter pylori neutrophil-activating protein (HP-NAP) is a virulence factor that attracts and activates neutrophils, and promotes their endothelial adhesion and the production of oxygen radicals and chemokines, including CXCL8, CCL3 and CCL4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Helicobacter Pylori HP-NAP
Applications ELISA
Classification Recombinant Monoclonal
Clone RM413
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide
Host Species Rabbit
Immunogen Proteins purified from Helicobacter pylori which correspond to the full length protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.