Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Helicobacter Pylori HP-NAP Recombinant Rabbit Monoclonal Antibody (RM414)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA544571
Description
This antibody reacts to Histone H4 symmetric dimethylated at Arginine 3 (R3me2s).
In H. pylori, the Helicobacter pylori neutrophil-activating protein (HP-NAP) is a virulence factor that attracts and activates neutrophils, and promotes their endothelial adhesion and the production of oxygen radicals and chemokines, including CXCL8, CCL3 and CCL4.
Specifications
Helicobacter Pylori HP-NAP | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide | |
Proteins purified from Helicobacter pylori which correspond to the full length protein. | |
100 μL | |
Primary | |
-20°C | |
Liquid |
ELISA | |
RM414 | |
Unconjugated | |
Rabbit | |
Protein A | |
RUO | |
Bacteria | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction