Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Helicobacter pylori Monoclonal Antibody (1B5)
GREENER_CHOICE

Catalog No. PIMA516300
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA516300 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA516300 Supplier Invitrogen™ Supplier No. MA516300
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Helicobacter pylori Monoclonal Antibody for ELISA, LF

Helicobacter Pylori (H. Pylori) is known to cause peptic ulcers and chronic gastritis in human. It is associated with duodenal ulcers and may be involved in development of adenocarcimona and low grade lymphoma of mucosa associated lymphoid tissue in the stomach. It is a microaerophilic gram-Negative bacterium, helix-shaped, and with many diverse strains.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Helicobacter pylori
Applications ELISA, Lateral Flow
Classification Monoclonal
Clone 1B5
Concentration 3.41 mg/mL
Conjugate Unconjugated
Formulation PBS with 15mM sodium azide; pH 7.5
Gene Alias H. pylori
Host Species Mouse
Immunogen Neutrophil-activating protein (NAP) bound to nitrocellulose, followed by NAP native. Mouse 124: CB6/F1-Generation (BALBc x C57BI).
Purification Method Protein G
Quantity 2 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.